Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-4-nitrobenzene-1-sulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A219541-100MG A219541-1G A219541-250MG A219541-5G
77097-918EA 80 USD InStock
77097-918 77097-920 77097-922 77097-924
3-Methoxy-4-nitrobenzene-1-sulfonyl chloride 95%
3-Methoxy-4-nitrobenzene-1-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₇H₆ClNO₅S
MDL Number: MFCD18398367
CAS Number: 1261646-30-3
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty