Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-cyclopentene-1-carboxylate 98%

Supplier: AMBEED, INC
Danger

A221318-100G A221318-1G A221318-25G A221318-5G
77097-960EA 479 USD InStock
77097-960 77097-962 77097-964 77097-966
Ethyl-3-cyclopentene-1-carboxylate 98%
Ethyl-3-cyclopentene-1-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂O₂
MDL Number: MFCD06410819
CAS Number: 21622-01-5
UN: 3272
ADR: 3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty