Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Bromopyridin-3-yl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone 98%

Supplier: AMBEED, INC
Warning

A223382-10MG A223382-50MG A223382-5MG A223382-100MG
77097-998EA 65 USD InStock
77097-998 77098-000 77098-002 77098-004 77685-605
(5-Bromopyridin-3-yl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone 98%
(5-Bromopyridin-3-yl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone
Precaución: for Research Use Only


Formula: C₁₅H₂₀BrN₃O
MDL Number: MFCD26936345
CAS Number: 1314241-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty