Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(chloromethyl)pyridine 96%

Supplier: AMBEED, INC
Danger

A225346-10G A225346-25G
77098-068EA 6 USD InStock
77098-068 77098-070 77098-072
2-Chloro-5-(chloromethyl)pyridine 96%
2-Chloro-5-(chloromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 37
MDL Number: MFCD00125366
CAS Number: 70258-18-3
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty