Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydro-2H-pyrano[2,3-c]pyridine-6-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A229772-100MG A229772-1G A229772-250MG
77098-196EA 88 USD InStock
77098-196 77098-198 77098-200
3,4-Dihydro-2H-pyrano[2,3-c]pyridine-6-carbaldehyde 95%
3,4-Dihydro-2H-pyrano[2,3-c]pyridine-6-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₉NO₂
MDL Number: MFCD18802894
CAS Number: 527681-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty