Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl isonicotinate 95%

Supplier: AMBEED, INC
Warning

A229993-1000G A229993-100G A229993-10G A229993-25G A229993-500G
77098-204EA 220 USD InStock
77098-204 77098-206 77098-208 77098-210 77098-212
Methyl isonicotinate 95%
Methyl isonicotinate
Precaución: for Research Use Only


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 207
Melting Pt: 17
MDL Number: MFCD00006427
CAS Number: 2459-09-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty