Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-norvaline 98%

Supplier: AMBEED, INC
Warning

A229996-1G A229996-25G A229996-5G A229996-100G
77098-214EA 10 USD InStock
77098-214 77098-216 77098-218 77685-628
Boc-L-norvaline 98%
Boc-L-norvaline
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₄
MDL Number: MFCD00037268
CAS Number: 53308-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty