Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-homocitrulline 97%

Supplier: AMBEED, INC
Warning

A237152-10G A237152-1G A237152-250MG A237152-25G A237152-5G
77098-362EA 166 USD InStock
77098-362 77098-364 77098-366 77098-368 77098-370
Fmoc-L-homocitrulline 97%
Fmoc-L-homocitrulline
Precaución: for Research Use Only


Formula: C₂₂H₂₅N₃O₅
MW: 411.458 g/mol
MDL Number: MFCD00237387
CAS Number: 201485-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty