Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Chloro-4'-fluoroacetophenone 97%

Supplier: AMBEED, INC
Warning

A239344-100G A239344-10G A239344-25G A239344-500G A239344-5G
77098-434EA 71 USD InStock
77098-434 77098-436 77098-438 77098-440 77098-442 77098-444
3'-Chloro-4'-fluoroacetophenone 97%
3'-Chloro-4'-fluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆ClFO
MW: 172.59
MDL Number: MFCD00042203
CAS Number: 2923-66-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty