Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Di(piperidin-1-yl)ethane-1,2-dithione 95%

Supplier: AMBEED, INC
Warning

A240262-100MG A240262-1G A240262-250MG
77098-450EA 143 USD InStock
77098-450 77098-452 77098-454
1,2-Di(piperidin-1-yl)ethane-1,2-dithione 95%
1,2-Di(piperidin-1-yl)ethane-1,2-dithione
Precaución: for Research Use Only


Formula: C₁₂H₂₀N₂S₂
MW: 256.43 g/mol
MDL Number: MFCD30533928
CAS Number: 24528-76-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty