Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(Bromomethyl)-3-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: AMBEED, INC
Danger

A242102-100MG A242102-1G A242102-250MG A242102-25G A242102-5G
77098-486EA 19 USD InStock
77098-486 77098-488 77098-490 77098-492 77098-494
2-(4-(Bromomethyl)-3-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-(Bromomethyl)-3-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₃H₁₇BBrClO₂
MDL Number: MFCD18783178
CAS Number: 1402238-25-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty