Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Difluoro-2-methoxyphenylacetonitrile 95%

Supplier: AMBEED, INC
Warning

A243555-100MG A243555-1G A243555-250MG A243555-5G
77098-532EA 6 USD InStock
77098-532 77098-534 77098-536 77098-538
4,5-Difluoro-2-methoxyphenylacetonitrile 95%
4,5-Difluoro-2-methoxyphenylacetonitrile
Precaución: for Research Use Only


Formula: C₉H₇F₂NO
MDL Number: MFCD06660349
CAS Number: 886761-74-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty