Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-hydroxynicotinic acid 97%

Supplier: AMBEED, INC
Warning

A243780-100G A243780-100MG A243780-10G A243780-1G A243780-250MG A243780-25G A243780-5G
77098-540EA 980 USD InStock
77098-540 77098-542 77098-544 77098-546 77098-548 77098-550 77098-552
2-Chloro-6-hydroxynicotinic acid 97%
2-Chloro-6-hydroxynicotinic acid
Precaución: for Research Use Only


Formula: C₆H₄ClNO₃
MDL Number: MFCD09757508
CAS Number: 38025-90-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty