Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Vanillylnonanamide [=Capsaicin (Synthetic)] 98%

Supplier: AMBEED, INC
Danger

A244101-100G A244101-10G A244101-1G A244101-5G
77098-558EA 223 USD InStock
77098-558 77098-560 77098-562 77098-564 77098-566
N-Vanillylnonanamide [=Capsaicin (Synthetic)] 98%
N-Vanillylnonanamide [=Capsaicin (Synthetic)]
Precaución: for Research Use Only


Formula: C₁₇H₂₇NO₃
MDL Number: MFCD00017286
CAS Number: 2444-46-4
UN: 3335
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty