Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A254199-100G A254199-10G A254199-1G A254199-25G A254199-500G
77098-720EA 78 USD InStock
77098-720 77098-722 77098-724 77098-726 77685-722
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 97%
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₅H₂₀ClNO₃
MDL Number: MFCD00012792
CAS Number: 52763-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty