Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitroquinoline 95%

Supplier: AMBEED, INC
Warning

A259426-100MG A259426-250MG
77098-794EA 15 USD InStock
77098-794 77098-796
3-Nitroquinoline 95%
3-Nitroquinoline
Precaución: for Research Use Only


Formula: C₉H₆N₂O₂
MDL Number: MFCD00956379
CAS Number: 17576-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty