Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[(Tetrahydro-2H-pyran-4-yl)methyl]hydrazine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A265089-100MG A265089-250MG
77098-912EA 99 USD InStock
77098-912 77098-914
[(Tetrahydro-2H-pyran-4-yl)methyl]hydrazine dihydrochloride 95%
[(Tetrahydro-2H-pyran-4-yl)methyl]hydrazine dihydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄N₂O·2ClH
MDL Number: MFCD19382069
CAS Number: 1315365-54-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty