Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Val-Ala-OH 95%

Supplier: AMBEED, INC
Warning

A266251-1G A266251-250MG A266251-5G
77098-942EA 20 USD InStock
77098-942 77098-944 77098-946
Boc-Val-Ala-OH 95%
Boc-Val-Ala-OH
Precaución: for Research Use Only


Formula: C₁₃H₂₄N₂O₅
MDL Number: MFCD22381612
CAS Number: 70396-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty