Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Harpagoside 98%

Supplier: AMBEED, INC
Warning

A268491-10MG A268491-1MG A268491-5MG
77098-964EA 41 USD InStock
77098-964 77098-966 77098-968
Harpagoside 98%
Harpagoside
Precaución: for Research Use Only


Formula: C₂₄H₃₀O₁₁
MW: 494.5 g/mol
MDL Number: MFCD00017415
CAS Number: 19210-12-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty