Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Phenylalanine benzyl ester-p-toluenesulfonate 98%

Supplier: AMBEED, INC
Warning

A270303-100G A270303-25G A270303-5G
77098-976EA 105 USD InStock
77098-976 77098-978 77098-980
D-Phenylalanine benzyl ester-p-toluenesulfonate 98%
D-Phenylalanine benzyl ester-p-toluenesulfonate
Precaución: for Research Use Only


Formula: C₂₃H₂₅NO₅S
MW: 427.52 g/mol
MDL Number: MFCD00066129
CAS Number: 28607-46-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty