Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butyl-3-methylimidazoliumhexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A272226-100G A272226-10G A272226-25G A272226-500G A272226-5G
77099-016EA 43 USD InStock
77099-016 77099-018 77099-020 77099-022 77099-024
1-Butyl-3-methylimidazoliumhexafluorophosphate 98%
1-Butyl-3-methylimidazoliumhexafluorophosphate
Precaución: for Research Use Only


Formula: C₈H₁₅F₆N₂P
MDL Number: MFCD03093295
CAS Number: 174501-64-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty