Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N6-Benzoyl-2'-fluoro-2'-deoxyadenosine 98+%

Supplier: AMBEED, INC
Warning

A274171-1G A274171-250MG A274171-5G
77099-052EA 13 USD InStock
77099-052 77099-054 77099-056
N6-Benzoyl-2'-fluoro-2'-deoxyadenosine 98+%
N6-Benzoyl-2'-fluoro-2'-deoxyadenosine
Precaución: for Research Use Only


Formula: C₁₇H₁₆FN₅O₄
MDL Number: MFCD00009628
CAS Number: 305808-19-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty