Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A277463-10G A277463-25G A277463-500G
77099-114EA 5 USD InStock
77099-114 77099-116 77099-118
4-Fluoro-3-nitroaniline 98%
4-Fluoro-3-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD00007833
CAS Number: 364-76-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty