Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromophenyl)acetonitrile 98%

Supplier: AMBEED, INC
Warning

A279735-100G A279735-10G A279735-25G A279735-500G A279735-5G
77099-150EA 40 USD InStock
77099-150 77099-152 77099-154 77099-156 77099-158
(3-Bromophenyl)acetonitrile 98%
(3-Bromophenyl)acetonitrile
Precaución: for Research Use Only


Formula: C₈H₆BrN
MDL Number: MFCD00001906
CAS Number: 31938-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty