Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tofacitinib 98%

Supplier: AMBEED, INC
Danger

A280750-1MG A280750-50MG A280750-5MG A280750-100MG A280750-1G
77099-170EA 8 USD InStock
77099-170 77099-172 77099-174 77685-792 77685-793
Tofacitinib 98%
Tofacitinib
Precaución: for Research Use Only


Formula: C₁₆H₂₀N₆O
MW: 312.4 g/mol
MDL Number: MFCD11035919
CAS Number: 477600-75-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty