Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluorophenol 98%

Supplier: AMBEED, INC
Danger

A282742-10G A282742-1G A282742-5G A282742-25G
77099-188EA 8 USD InStock
77099-188 77099-190 77099-192 77685-802
2-Chloro-6-fluorophenol 98%
2-Chloro-6-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 199
Melting Pt: 65
MDL Number: MFCD01631574
CAS Number: 2040-90-6
UN: 1759
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty