Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Pyridyl)acetic acid ethyl ester 98%

Supplier: AMBEED, INC
Warning

A284002-100G A284002-1G A284002-5G
77099-212EA 920 USD InStock
77099-212 77099-214 77099-216
(4-Pyridyl)acetic acid ethyl ester 98%
(4-Pyridyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.19
MDL Number: MFCD00051830
CAS Number: 54401-85-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty