Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluorophenyl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate 95%

Supplier: AMBEED, INC
Warning

A285266-100MG A285266-1G A285266-250MG
77099-240EA 43 USD InStock
77099-240 77099-242 77099-244
Perfluorophenyl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate 95%
Perfluorophenyl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate
Precaución: for Research Use Only


Formula: C₁₆H₁₂F₅NO₄
MDL Number: MFCD18916982
CAS Number: 692739-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty