Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzyl alcohol 95%

Supplier: AMBEED, INC
Warning

A290769-1000G A290769-10G A290769-25G A290769-500G
77099-308EA 288 USD InStock
77099-308 77099-310 77099-312 77099-314 77099-316
4-Fluorobenzyl alcohol 95%
4-Fluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₇FO
MDL Number: MFCD00004651
CAS Number: 459-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty