Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorophenyl trifluoromethanesulfonate 98+%

Supplier: AMBEED, INC
Warning

A292990-1G A292990-250MG A292990-25G A292990-5G
77099-362EA 29 USD InStock
77099-362 77099-364 77099-366 77099-368
2-Chlorophenyl trifluoromethanesulfonate 98+%
2-Chlorophenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₇H₄ClF₃O₃S
MDL Number: MFCD07784322
CAS Number: 66107-36-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty