Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-acetamido-2-chloro-5-iodobenzoate 95%

Supplier: AMBEED, INC
Warning

A296210-1G A296210-250MG A296210-5G
77099-426EA 39 USD InStock
77099-426 77099-428 77099-430
Methyl 4-acetamido-2-chloro-5-iodobenzoate 95%
Methyl 4-acetamido-2-chloro-5-iodobenzoate
Precaución: for Research Use Only


Formula: C₁₀H₉ClINO₃
MW: 353.54 g/mol
MDL Number: MFCD09260862
CAS Number: 672292-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty