Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Phenylamino)benzaldehyde 95%

Supplier: AMBEED, INC
Warning

A298244-100MG A298244-1G A298244-250MG A298244-25G A298244-5G
77099-470EA 5 USD InStock
77099-470 77099-472 77099-474 77099-476 77099-478
4-(Phenylamino)benzaldehyde 95%
4-(Phenylamino)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₃H₁₁NO
MDL Number: MFCD03950803
CAS Number: 100727-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty