Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenazine-1,6-dicarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A301402-100MG A301402-250MG
77099-536EA 472 USD InStock
77099-536 77099-538
Phenazine-1,6-dicarboxylic acid 95%
Phenazine-1,6-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₈N₂O₄
MW: 268.22 g/mol
MDL Number: MFCD00182804
CAS Number: 23462-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty