Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-3-hydroxypropanamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A305643-10G A305643-1G A305643-25G A305643-5G
77099-636EA 55 USD InStock
77099-636 77099-638 77099-640 77099-642
(R)-2-Amino-3-hydroxypropanamide hydrochloride 95%
(R)-2-Amino-3-hydroxypropanamide hydrochloride
Precaución: for Research Use Only


Formula: C₃H₉ClN₂O₂
MDL Number: MFCD00050702
CAS Number: 122702-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty