Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-hydroxy-3-nitro-4-picoline 98%

Supplier: AMBEED, INC
Warning

A314414-100G A314414-10G A314414-1G A314414-250MG A314414-25G A314414-5G
77099-748EA 151 USD InStock
77099-748 77099-750 77099-752 77099-754 77099-756 77099-758
5-Bromo-2-hydroxy-3-nitro-4-picoline 98%
5-Bromo-2-hydroxy-3-nitro-4-picoline
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₃
MW: 233.0200 g/mol
MDL Number: MFCD06659520
CAS Number: 228410-90-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty