Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-(4-Pyridyl)-2,2':6',2''-terpyridine 97%

Supplier: AMBEED, INC
Warning

A316429-250MG A316429-100MG A316429-1G A316429-25G
77099-778EA 12 USD InStock
77099-778 77685-896 77685-897 77685-898
4'-(4-Pyridyl)-2,2':6',2''-terpyridine 97%
4'-(4-Pyridyl)-2,2':6',2''-terpyridine
Precaución: for Research Use Only


Formula: C₂₀H₁₄N₄
MW: 310.36 g/mol
MDL Number: MFCD00932037
CAS Number: 112881-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty