Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-fluorophenol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A319188-1G A319188-250MG A319188-5G
77099-818EA 13 USD InStock
77099-818 77099-820 77099-822
4-Amino-2-fluorophenol hydrochloride 95%
4-Amino-2-fluorophenol hydrochloride


Formula: C₆H₇ClFNO
MDL Number: MFCD28291809
CAS Number: 1341216-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty