Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(Diethylamino)methyl]benzonitrile 98%

Supplier: AMBEED, INC
Warning

A324568-100MG A324568-1G A324568-250MG A324568-5G
77099-888EA 8 USD InStock
77099-888 77099-890 77099-892 77099-894
4-[(Diethylamino)methyl]benzonitrile 98%
4-[(Diethylamino)methyl]benzonitrile
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂
MDL Number: MFCD09948026
CAS Number: 69293-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty