Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Fluoro-3'-(trifluoromethyl)acetophenone 98%

Supplier: AMBEED, INC
Warning

A335297-10G A335297-1G
77106-560EA 30 USD InStock
77106-560 77106-562
4'-Fluoro-3'-(trifluoromethyl)acetophenone 98%
4'-Fluoro-3'-(trifluoromethyl)acetophenone
Precaución: for Research Use Only


Formula: C₉H₆F₄O
MDL Number: MFCD00061252
CAS Number: 208173-24-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty