Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Pyridyl)propionic acid 98%

Supplier: AMBEED, INC
Warning

A344219-1G A344219-25G A344219-5G
77106-696EA 10 USD InStock
77106-696 77106-698 77106-700
3-(3-Pyridyl)propionic acid 98%
3-(3-Pyridyl)propionic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MDL Number: MFCD00274197
CAS Number: 3724-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty