Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzo[b]thiophen-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: AMBEED, INC
Warning

A349294-100MG A349294-1G A349294-250MG
77106-756EA 16 USD InStock
77106-756 77106-758 77106-760
2-(Benzo[b]thiophen-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(Benzo[b]thiophen-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₁₇BO₂S
MDL Number: MFCD13619876
CAS Number: 1000160-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty