Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Acetamidobutanoic acid 97%

Supplier: AMBEED, INC
Warning

A349732-100G A349732-1G A349732-5G A349732-25G
77106-776EA 163 USD InStock
77106-776 77106-778 77106-780 77106-782 77685-993
(R)-2-Acetamidobutanoic acid 97%
(R)-2-Acetamidobutanoic acid
Precaución: for Research Use Only


Formula: C₆H₁₁NO₃
MW: 145.15 g/mol
MDL Number: MFCD01862370
CAS Number: 34271-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty