Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Mandelamide 95%

Supplier: AMBEED, INC
Warning

A353553-100G A353553-10G A353553-1G A353553-25G A353553-5G
77106-814EA 186 USD InStock
77106-814 77106-816 77106-818 77106-820 77106-822
DL-Mandelamide 95%
DL-Mandelamide
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00025495
CAS Number: 4410-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty