Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-benzenesulfonamidobenzoate 95%

Supplier: AMBEED, INC
Warning

A361243-10G A361243-1G A361243-5G
77106-918EA 364 USD InStock
77106-918 77106-920 77106-922
Methyl 3-benzenesulfonamidobenzoate 95%
Methyl 3-benzenesulfonamidobenzoate
Precaución: for Research Use Only


Formula: C₁₄H₁₃NO₄S
MDL Number: MFCD00121318
CAS Number: 107922-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty