Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluoro-1-(4-nitrophenyl)butane-1,3-dione 98%

Supplier: AMBEED, INC
Warning

A363395-100MG A363395-250MG A363395-5G
77106-936EA 18 USD InStock
77106-936 77106-938 77106-940
4,4,4-Trifluoro-1-(4-nitrophenyl)butane-1,3-dione 98%
4,4,4-Trifluoro-1-(4-nitrophenyl)butane-1,3-dione
Precaución: for Research Use Only


Formula: C₁₀H₆F₃NO₄
MW: 261.15 g/mol
MDL Number: MFCD00440667
CAS Number: 35999-53-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty