Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-amino-3,5,6-trichloropicolinate 95%

Supplier: AMBEED, INC
Warning

A365106-100MG A365106-1G A365106-250MG A365106-5G
77106-956EA 5 USD InStock
77106-956 77106-958 77106-960 77106-962
Ethyl 4-amino-3,5,6-trichloropicolinate 95%
Ethyl 4-amino-3,5,6-trichloropicolinate
Precaución: for Research Use Only


Formula: C₈H₇Cl₃N₂O₂
MDL Number: MFCD00183796
CAS Number: 91867-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty