Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3,4-Dichloro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: AMBEED, INC
Warning

A369432-100MG A369432-1G A369432-250MG
77101-552EA 12 USD InStock
77101-552 77101-554 77101-556
2-(3,4-Dichloro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3,4-Dichloro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₃H₁₇BCl₂O₃
MDL Number: MFCD19237221
CAS Number: 1701449-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty