Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-8-fluoronaphthalene 98%

Supplier: AMBEED, INC
Warning

A371709-100MG A371709-1G A371709-250MG A371709-5G
77101-588EA 12 USD InStock
77101-588 77101-590 77101-592 77101-594
1-Bromo-8-fluoronaphthalene 98%
1-Bromo-8-fluoronaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₆BrF
MW: 225.05 g/mol
MDL Number: MFCD07368856
CAS Number: 33718-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty