Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Benzyl tert-butyl (3-hydroxypropane-1,2-diyl)dicarbamate 97%

Supplier: AMBEED, INC
Warning

A383579-100MG A383579-1G A383579-250MG
77101-722EA 22 USD InStock
77101-722 77101-724 77101-726
(R)-Benzyl tert-butyl (3-hydroxypropane-1,2-diyl)dicarbamate 97%
(R)-Benzyl tert-butyl (3-hydroxypropane-1,2-diyl)dicarbamate
Precaución: for Research Use Only


Formula: C₁₆H₂₃N₂O₅-
MDL Number: MFCD06798156
CAS Number: 1263045-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty