Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-fluoro-2-iodobenzoate 95%

Supplier: AMBEED, INC
Warning

A390679-1G A390679-250MG A390679-5G
77101-766EA 23 USD InStock
77101-766 77101-768 77101-770
Methyl 3-fluoro-2-iodobenzoate 95%
Methyl 3-fluoro-2-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₆FIO₂
MW: 280.035 g/mol
MDL Number: MFCD18399305
CAS Number: 393-78-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty